siRNA Details
| Gene Symbol: A930007I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015567.2 | 2 | 129 | |||
| GenBank mRNA | AK020826.1 | - | 177 | |||
| AK043087.1 | - | 137 | ||||
| AK137192.1 | - | 2 | 131 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930007I19Rik| Gene Symbol: A930007I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015567.2 | 2 | 129 | |||
| GenBank mRNA | AK020826.1 | - | 177 | |||
| AK043087.1 | - | 137 | ||||
| AK137192.1 | - | 2 | 131 | |||