siRNA Details
Gene Symbol: Gm5627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033301.1 | 4 | 879 | |||
GenBank mRNA | AK041874.1 | - | 4 | 882 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033301.1 | 4 | 879 | |||
GenBank mRNA | AK041874.1 | - | 4 | 882 |