siRNA Details
Gene Symbol: Rian | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028261.1 | 21 | 3062 | |||
GenBank mRNA | AF498294.1 | - | 11 | 1830 | ||
AK017440.1 | - | 11 | 1791 | |||
EU434919.1 | - | 26 | 2701 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ445470.1 | - | 4 | 653 | Chr. 12 - 109655886 - 109661307 [+] on Build GRCm38 |
