siRNA Details
| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028261.1 | 21 | 2970 | |||
| GenBank mRNA | AF498294.1 | - | 11 | 1738 | ||
| AK017440.1 | - | 11 | 1699 | |||
| EU434919.1 | - | 26 | 2609 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rian| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028261.1 | 21 | 2970 | |||
| GenBank mRNA | AF498294.1 | - | 11 | 1738 | ||
| AK017440.1 | - | 11 | 1699 | |||
| EU434919.1 | - | 26 | 2609 | |||