siRNA Details
| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028261.1 | 11 | 1178 | |||
| GenBank mRNA | AF498301.1 | - | 8 | 775 | ||
| AF498302.1 | - | 8 | 824 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rian| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028261.1 | 11 | 1178 | |||
| GenBank mRNA | AF498301.1 | - | 8 | 775 | ||
| AF498302.1 | - | 8 | 824 | |||