siRNA Details
| Gene Symbol: Rian | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_028261.1 | 11 | 1673 | |||
| GenBank mRNA | AB063319.1 | - | LIT1623 Rian |
1 | 432 | |
| AF498294.1 | - | 1 | 450 | |||
| AK017440.1 | - | 1 | 402 | |||

Rian
Genomic Map