siRNA Details
| Gene Symbol: Gm15698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003564.1 | 1 | 156 | |||
| GenBank mRNA | BC099439.1 | - | 1 | 156 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15698| Gene Symbol: Gm15698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003564.1 | 1 | 156 | |||
| GenBank mRNA | BC099439.1 | - | 1 | 156 | ||