siRNA Details
| Gene Symbol: A930015D03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015618.2 | 2 | 579 | |||
| GenBank mRNA | AK020860.1 | - | 2 | 580 | ||
| AK078380.1 | - | 2 | 629 | |||
| AK142942.1 | - | 2 | 575 | |||
| BC055918.1 | - | 2 | 557 | |||
| Gene Symbol: BC023719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC023719.1 | - | 2 | 500 | ||

A930015D03Rik
Genomic Map