siRNA Details
Gene Symbol: Zfp672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028331.2 | 2 | 143 | |||
NR_046280.1 | 2 | 143 | ||||
GenBank mRNA | AK008471.1 | - | 2 | 119 | ||
AK019676.1 | - | 2 | 93 | |||
AK029711.1 | - | 2 | 311 | |||
AK085166.1 | - | 2 | 103 | |||
BB615378.1 | - | 2 | 304 | |||
BC034706.1 | - | 2 | 90 | 3582315 | ||
BG091612.1 | - | 2 | 104 | |||
BU148968.1 | - | 2 | 75 | |||
BY017375.1 | - | 2 | 124 | |||
BY715308.1 | - | 2 | 93 | |||
CA321162.1 | - | 2 | 76 | |||
CV783882.1 | - | 2 | 92 | |||
DV654279.1 | - | 2 | 177 |
