siRNA Details
| Gene Symbol: Bambi-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027919.1 | 1 | 378 | |||
| GenBank mRNA | AK040009.1 | - | 1 | 322 | ||
| AK041875.1 | - | 1 | 324 | |||
| AK088217.1 | - | 1 | 316 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bambi-ps1| Gene Symbol: Bambi-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027919.1 | 1 | 378 | |||
| GenBank mRNA | AK040009.1 | - | 1 | 322 | ||
| AK041875.1 | - | 1 | 324 | |||
| AK088217.1 | - | 1 | 316 | |||