siRNA Details
| Gene Symbol: 4930442L01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015596.1 | 2 | 67 | |||
| GenBank mRNA | AK006795.1 | - | 1 | 242 | ||
| AK006978.1 | - | 2 | 47 | |||
| AK015364.1 | - | 2 | 67 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930442L01Rik| Gene Symbol: 4930442L01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015596.1 | 2 | 67 | |||
| GenBank mRNA | AK006795.1 | - | 1 | 242 | ||
| AK006978.1 | - | 2 | 47 | |||
| AK015364.1 | - | 2 | 67 | |||