siRNA Details
| Gene Symbol: PTGER4P2-CDK2AP2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024496.1 | 2 | 3384 | |||
| NR_135010.1 | 3 | 2803 | ||||
| GenBank mRNA | BC068605.1 | - | 3 | 2801 | ||
| Gene Symbol: FLJ37786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK095105.1 | - | 1 | 1854 | ||
| Gene Symbol: LOC105371215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011510253.2 | XP_011508555.1 | 2 | 1383 | ||
| Gene Symbol: LOC107985527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017003061.1 | XP_016858550.1 | 2 | 1148 | ||

PTGER4P2-CDK2AP2P2
Genomic Map