siRNA Details
| Gene Symbol: PTGER4P2-CDK2AP2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024496.1 | 2 | 1133 | |||
| NR_135010.1 | 3 | 552 | ||||
| GenBank mRNA | BC042539.1 | - | 2 | 1133 | ||
| BC068605.1 | - | 3 | 551 | |||
| Gene Symbol: LOC105379526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | X97876.1 | - | 3 | 1424 | ||

PTGER4P2-CDK2AP2P2
Genomic Map