siRNA Details
| Gene Symbol: LOC100130872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024569.1 | 4 | 3275 | |||
| GenBank mRNA | AK122772.1 | - | 1 | 327 | ||
| BC030552.1 | - | 1 | 265 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100130872| Gene Symbol: LOC100130872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024569.1 | 4 | 3275 | |||
| GenBank mRNA | AK122772.1 | - | 1 | 327 | ||
| BC030552.1 | - | 1 | 265 | |||