siRNA Details
Gene Symbol: LINC00305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027245.1 | 3, 4 | 483 | |||
GenBank mRNA | AY336744.1 | - | 2, 3 | 246 | ||
BC029565.1 | - | 3, 4 | 487 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027245.1 | 3, 4 | 483 | |||
GenBank mRNA | AY336744.1 | - | 2, 3 | 246 | ||
BC029565.1 | - | 3, 4 | 487 |