siRNA Details
| Gene Symbol: LINC00305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027245.1 | 4 | 681 | |||
| GenBank mRNA | BC029565.1 | - | 4 | 685 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00305| Gene Symbol: LINC00305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027245.1 | 4 | 681 | |||
| GenBank mRNA | BC029565.1 | - | 4 | 685 | ||