siRNA Details
Gene Symbol: LINC00526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026849.1 | 1 | 326 | |||
GenBank mRNA | AK292743.1 | - | 1 | 903 | ||
BC010538.1 | - | 1 | 326 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026849.1 | 1 | 326 | |||
GenBank mRNA | AK292743.1 | - | 1 | 903 | ||
BC010538.1 | - | 1 | 326 |