siRNA Details
| Gene Symbol: CCDC162P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028595.1 | 4 | 531 | |||
| GenBank mRNA | BC107113.2 | - | 5 | 671 | 40022743 | |
| BC107114.2 | - | 4 | 531 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC162P| Gene Symbol: CCDC162P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028595.1 | 4 | 531 | |||
| GenBank mRNA | BC107113.2 | - | 5 | 671 | 40022743 | |
| BC107114.2 | - | 4 | 531 | |||