siRNA Details
| Gene Symbol: VLDLR-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015375.2 | 4 | 547 | |||
| GenBank mRNA | AF424541.1 | - | 8 | 1012 | ||
| BC004474.2 | - | 4 | 547 | |||
| BE086096.1 | - | 4 | 343 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

VLDLR-AS1| Gene Symbol: VLDLR-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015375.2 | 4 | 547 | |||
| GenBank mRNA | AF424541.1 | - | 8 | 1012 | ||
| BC004474.2 | - | 4 | 547 | |||
| BE086096.1 | - | 4 | 343 | |||