siRNA Details
| Gene Symbol: KATNBL1P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003954.1 | 1 | 1720 | |||
| GenBank mRNA | BC067801.1 | - | 1 | 1720 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KATNBL1P6| Gene Symbol: KATNBL1P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003954.1 | 1 | 1720 | |||
| GenBank mRNA | BC067801.1 | - | 1 | 1720 | ||