siRNA Details
| Gene Symbol: SHROOM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027121.1 | 9, 10 | 5501 | |||
| GenBank mRNA | AB033028.1 | - | 9, 10 | 5516 | ||
| BC151240.1 | - | 9, 10 | 5527 | 8860513 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SHROOM4| Gene Symbol: SHROOM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027121.1 | 9, 10 | 5501 | |||
| GenBank mRNA | AB033028.1 | - | 9, 10 | 5516 | ||
| BC151240.1 | - | 9, 10 | 5527 | 8860513 | ||