siRNA Details
Gene Symbol: LOC90246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026954.1 | 1 | 1787 | |||
GenBank mRNA | AK023635.1 | - | 1 | 1479 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC90246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026954.1 | 1 | 1787 | |||
GenBank mRNA | AK023635.1 | - | 1 | 1479 |