siRNA Details
| Gene Symbol: OLMALINC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026762.1 | 1 | 265 | |||
| GenBank mRNA | BC024924.1 | - | 1 | 226 | ||
| BC045787.1 | - | 1 | 244 | |||
| CB159470.1 | - | 1 | 292 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OLMALINC| Gene Symbol: OLMALINC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026762.1 | 1 | 265 | |||
| GenBank mRNA | BC024924.1 | - | 1 | 226 | ||
| BC045787.1 | - | 1 | 244 | |||
| CB159470.1 | - | 1 | 292 | |||