siRNA Details
| Gene Symbol: OLMALINC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026762.1 | 2 | 502 | |||
| GenBank mRNA | AK023036.1 | - | 2 | 235 | ||
| BC024924.1 | - | 2 | 463 | |||
| BC045787.1 | - | 2 | 481 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OLMALINC| Gene Symbol: OLMALINC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026762.1 | 2 | 502 | |||
| GenBank mRNA | AK023036.1 | - | 2 | 235 | ||
| BC024924.1 | - | 2 | 463 | |||
| BC045787.1 | - | 2 | 481 | |||