siRNA Details
| Gene Symbol: COG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026745.1 | 1, 2 | 252 | |||
| GenBank mRNA | AF116827.1 | - | 1, 2 | 135 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AL599284.1 | - | 1, 2 | 156 | Chr. 13 - 39655724 - 39677494 [+] on Build GRCh38 | |

COG6
Genomic Map