siRNA Details
| Gene Symbol: ZNF674-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015378.3 | 2 | 874 | |||
| GenBank mRNA | AK056314.1 | - | 2 | 845 | ||
| BC034616.1 | - | 2 | 871 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM547124.1 | - | 2 | 834 | Chr. X - 46545552 - 46547418 [+] on Build GRCh38 | |

ZNF674-AS1
Genomic Map