siRNA Details
| Gene Symbol: ZNF674-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015378.3 | 2 | 1810 | |||
| GenBank mRNA | AK056314.1 | - | 2 | 1779 | ||
| BC034616.1 | - | 2 | 1806 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF674-AS1| Gene Symbol: ZNF674-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015378.3 | 2 | 1810 | |||
| GenBank mRNA | AK056314.1 | - | 2 | 1779 | ||
| BC034616.1 | - | 2 | 1806 | |||