siRNA Details
| Gene Symbol: FAM224B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002160.1 | 3 | 446 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM224B| Gene Symbol: FAM224B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002160.1 | 3 | 446 | |||