siRNA Details
| Gene Symbol: EPB41L4A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015370.2 | 3 | 1109 | |||
| GenBank mRNA | DA266578.1 | - | 3 | 477 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPB41L4A-AS1| Gene Symbol: EPB41L4A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015370.2 | 3 | 1109 | |||
| GenBank mRNA | DA266578.1 | - | 3 | 477 | ||