siRNA Details
| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026938.2 | 16 | 3247 | |||
| GenBank mRNA | AK126092.1 | - | 2 | 511 | ||
| AL833650.1 | - | 7 | 1137 | |||
| BX537595.1 | - | 13 | 4003 | |||
| BX648193.1 | - | 16 | 3243 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADCY10P1| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026938.2 | 16 | 3247 | |||
| GenBank mRNA | AK126092.1 | - | 2 | 511 | ||
| AL833650.1 | - | 7 | 1137 | |||
| BX537595.1 | - | 13 | 4003 | |||
| BX648193.1 | - | 16 | 3243 | |||