siRNA Details
| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026938.2 | 8 | 1733 | |||
| GenBank mRNA | BX537595.1 | - | 7 | 2034 | ||
| BX648193.1 | - | 8 | 1729 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADCY10P1| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026938.2 | 8 | 1733 | |||
| GenBank mRNA | BX537595.1 | - | 7 | 2034 | ||
| BX648193.1 | - | 8 | 1729 | |||