siRNA Details
| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026938.2 | 5 | 931 | |||
| GenBank mRNA | BX537595.1 | - | 4 | 1232 | ||
| BX648193.1 | - | 5 | 926 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADCY10P1| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026938.2 | 5 | 931 | |||
| GenBank mRNA | BX537595.1 | - | 4 | 1232 | ||
| BX648193.1 | - | 5 | 926 | |||