siRNA Details
| Gene Symbol: ADCY10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026938.2 | 23 | 4433 | |||
| GenBank mRNA | AK126092.1 | - | 9 | 3727 | ||
| AK128294.1 | - | 5 | 2907 | |||
| AL833650.1 | - | 12 | 1972 | |||
| BX537595.1 | - | 23 | 5685 | |||
| BX648193.1 | - | 23 | 4429 | |||

ADCY10P1
Genomic Map