siRNA Details
| Gene Symbol: LINC00638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024396.1 | 1 | 2140 | |||
| GenBank mRNA | AK095404.1 | - | 1 | 2140 | ||
| AK292179.1 | - | 1 | 1404 | |||
| BC015579.2 | - | 1 | 1402 | |||
| BC041036.1 | - | 1 | 1402 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00638| Gene Symbol: LINC00638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024396.1 | 1 | 2140 | |||
| GenBank mRNA | AK095404.1 | - | 1 | 2140 | ||
| AK292179.1 | - | 1 | 1404 | |||
| BC015579.2 | - | 1 | 1402 | |||
| BC041036.1 | - | 1 | 1402 | |||