siRNA Details
| Gene Symbol: RPL23AP64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003040.2 | 1 | 268 | |||
| GenBank mRNA | BC017930.1 | - | 1 | 265 | ||
| BC060042.1 | - | 1 | 268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL23AP64| Gene Symbol: RPL23AP64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003040.2 | 1 | 268 | |||
| GenBank mRNA | BC017930.1 | - | 1 | 265 | ||
| BC060042.1 | - | 1 | 268 | |||