siRNA Details
| Gene Symbol: TPT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024458.1 | 11 | 1221 | |||
| GenBank mRNA | AF318337.1 | - | 11 | 1221 | ||
| AK124332.1 | - | 8 | 1726 | |||
| AK127569.1 | - | 3 | 2572 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPT1-AS1| Gene Symbol: TPT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024458.1 | 11 | 1221 | |||
| GenBank mRNA | AF318337.1 | - | 11 | 1221 | ||
| AK124332.1 | - | 8 | 1726 | |||
| AK127569.1 | - | 3 | 2572 | |||