siRNA Details
| Gene Symbol: DNM1P46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003260.1 | 5 | 2999 | |||
| GenBank mRNA | AK127834.1 | - | 5 | 2999 | ||
| AK302717.1 | - | 5 | 2800 | |||
| AL833912.1 | - | 4 | 2854 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK125787.1 | - | 5 | 3622 | Chr. 15 - 99790781 - 99801688 [-] on Build GRCh38 | |

DNM1P46
Genomic Map