siRNA Details
Gene Symbol: ZNF529 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027239.1 | 3 | 2223 | |||
GenBank mRNA | AK091072.1 | - | 2 | 2298 | ||
AK094618.1 | - | 3 | 2209 | |||
AL109722.1 | - | 1 | 1234 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF529 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027239.1 | 3 | 2223 | |||
GenBank mRNA | AK091072.1 | - | 2 | 2298 | ||
AK094618.1 | - | 3 | 2209 | |||
AL109722.1 | - | 1 | 1234 |