siRNA Details
| Gene Symbol: ZNF529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027239.1 | 3 | 2325 | |||
| GenBank mRNA | AK091072.1 | - | 2 | 2400 | ||
| AK094618.1 | - | 3 | 2311 | |||
| AL109722.1 | - | 1 | 1336 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF529| Gene Symbol: ZNF529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027239.1 | 3 | 2325 | |||
| GenBank mRNA | AK091072.1 | - | 2 | 2400 | ||
| AK094618.1 | - | 3 | 2311 | |||
| AL109722.1 | - | 1 | 1336 | |||