siRNA Details
| Gene Symbol: ZNF529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027239.1 | 3 | 2151 | |||
| GenBank mRNA | AK091072.1 | - | 2 | 2226 | ||
| AK094618.1 | - | 3 | 2137 | |||
| AL109722.1 | - | 1 | 1162 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF529| Gene Symbol: ZNF529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027239.1 | 3 | 2151 | |||
| GenBank mRNA | AK091072.1 | - | 2 | 2226 | ||
| AK094618.1 | - | 3 | 2137 | |||
| AL109722.1 | - | 1 | 1162 | |||