siRNA Details
| Gene Symbol: AOX3P-AOX2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135011.1 | 28, 29 | 3854 | |||
| NR_135012.1 | 27, 28 | 3347 | ||||
| GenBank mRNA | JX431285.1 | - | 27, 28 | 3154 | ||
| JX431286.1 | - | 28, 29 | 3661 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AOX3P-AOX2P| Gene Symbol: AOX3P-AOX2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135011.1 | 28, 29 | 3854 | |||
| NR_135012.1 | 27, 28 | 3347 | ||||
| GenBank mRNA | JX431285.1 | - | 27, 28 | 3154 | ||
| JX431286.1 | - | 28, 29 | 3661 | |||