siRNA Details
| Gene Symbol: MGC15885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026897.1 | 3 | 1388 | |||
| GenBank mRNA | BC014404.1 | - | 3 | 1510 | ||
| BC131619.1 | - | 3 | 1287 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC15885| Gene Symbol: MGC15885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026897.1 | 3 | 1388 | |||
| GenBank mRNA | BC014404.1 | - | 3 | 1510 | ||
| BC131619.1 | - | 3 | 1287 | |||