siRNA Details
Gene Symbol: ZSCAN12P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024063.2 | 2 | 2870 | |||
GenBank mRNA | AK127188.1 | - | 1 | 392 | ||
BC040524.1 | - | 2 | 2526 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZSCAN12P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024063.2 | 2 | 2870 | |||
GenBank mRNA | AK127188.1 | - | 1 | 392 | ||
BC040524.1 | - | 2 | 2526 |