siRNA Details
Gene Symbol: ZSCAN12P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024063.2 | 2 | 1586 | |||
GenBank mRNA | AK055113.1 | - | 2 | 1248 | ||
BC040524.1 | - | 2 | 1242 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZSCAN12P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024063.2 | 2 | 1586 | |||
GenBank mRNA | AK055113.1 | - | 2 | 1248 | ||
BC040524.1 | - | 2 | 1242 |