siRNA Details
| Gene Symbol: LOC115110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037844.2 | 5 | 2979 | |||
| GenBank mRNA | AK128532.1 | - | 5 | 2979 | ||
| AL359943.1 | - | 1 | 2032 | |||
| CR749717.1 | - | 1 | 2032 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG470374.1 | - | 1 | 378 | Chr. 1 - 2551781 - 2552494 [-] on Build GRCh38 | |

LOC115110
Genomic Map