siRNA Details
| Gene Symbol: LOC115110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037844.2 | 5 | 2856 | |||
| GenBank mRNA | AK128532.1 | - | 5 | 2856 | ||
| AL359943.1 | - | 1 | 1909 | |||
| CR749717.1 | - | 1 | 1909 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC115110| Gene Symbol: LOC115110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037844.2 | 5 | 2856 | |||
| GenBank mRNA | AK128532.1 | - | 5 | 2856 | ||
| AL359943.1 | - | 1 | 1909 | |||
| CR749717.1 | - | 1 | 1909 | |||