siRNA Details
| Gene Symbol: MIR1302-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_031634.1 | 1 | 47 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR1302-5| Gene Symbol: MIR1302-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_031634.1 | 1 | 47 | |||