siRNA Details
Gene Symbol: AACSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024035.2 | 11 | 2803 | |||
NR_135095.1 | 11 | 2806 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AACSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024035.2 | 11 | 2803 | |||
NR_135095.1 | 11 | 2806 |