siRNA Details
| Gene Symbol: BRD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317951.1 | NP_001304880.1 | 4 | 648 | ||
| NR_134296.1 | 4 | 865 | ||||
| GenBank mRNA | AK023503.1 | - | 4 | 626 | ||
| AK024392.1 | - | 1 | 364 | |||
| AK301987.1 | - | 4 | 480 | |||
| BC007217.1 | - | 3 | 372 | |||
| BC041590.1 | - | 2 | 215 | 5428011 | ||
| BX648703.1 | - | 4 | 867 | |||

BRD9
Genomic Map