siRNA Details
| Gene Symbol: BRD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317951.1 | NP_001304880.1 | 6, 7 | 1012 | ||
| XM_006714494.3 | XP_006714557.1 | 6, 7 | 872 | |||
| XM_017009780.1 | XP_016865269.1 | 6, 7 | 872 | |||
| GenBank mRNA | AK023503.1 | - | 6, 7 | 990 | ||

BRD9
Genomic Map